Learn More
World Precision Instrument Carbon, 18 AWG, green
SDP
Catalog No. 502538885
Encompass
Carbon filament wire is made of 3,000 microscopic carbon strands in a green PVC jacket. 45.8 ohms/foot.
Catalog No. 50-253-8885 Supplier World Precision Instrument Supplier No. CPVC4050G
Carbon filament wire is made of 3,000 microscopic carbon strands in a green PVC jacket. 45.8 ohms/foot.