Learn More
World Precision Instrument Carbon, 18 AWG, white
SDP
Catalog No. 502538883
Encompass
Carbon filament wire is made of 3,000 microscopic carbon strands in a white PVC jacket. 45.8 ohms/foot.
Catalog No. 50-253-8883 Supplier World Precision Instrument Supplier No. CPVC4050W
Carbon filament wire is made of 3,000 microscopic carbon strands in a white PVC jacket. 45.8 ohms/foot.