Learn More
World Precision Instrument Carbon, 18 AWG, yellow
SDP
Supplier:  World Precision Instrument CPVC4050Y
Encompass
Carbon filament wire is made of 3,000 microscopic carbon strands in a yellow PVC jacket. 45.8 ohms/foot.
Catalog No. 50-253-8882