Learn More
Electron Microscopy Sciences Carbon Adhesive Tabs, 9 mm
SDP
Catalog No. 5025104
Encompass_Preferred
Two sides of thick conductive adhesive (45 micron on each side) with the center conductive core film (35 micron). Firm, smooth surface SEM including GSR. 125 um thick. No gas out-gassing. Traces of Si, Sb, S, Fe, Mg, Na.
Diameter: 9 mm
Catalog No. 50-251-04 Supplier Electron Microscopy Sciences Supplier No. 7782509
Two sides of thick conductive adhesive (45 micron on each side) with the center conductive core film (35 micron). Firm, smooth surface SEM including GSR. 125 um thick. No gas out-gassing. Traces of Si, Sb, S, Fe, Mg, Na.
Diameter: 9 mm