Learn More
National Safety Apparel™ Carbon Armour Double Layer FR Balaclava
Shop All National Safety Apparel Products
Description
- OPF/Para-Aramid Blend
- Mid-weight
- Double layer hood
Specifications
Specifications
| Size | Medium |
| Material | OPF/Para-Aramid Blend |
| Color | Olive Green |
| Seam Type | Stitched |
| Arc Flash Rating | 42 cal/cm² |
| Certifications/Compliance | CAT 4, NFPA 70 E, ASTM F1506, OSHA 1910.269, CSA Z462 |