Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ High Heat Knit Sleeve
SDP

Catalog No. 17355096 Shop All National Safety Apparel Products
Encompass_Preferred
Change view
Click to view available options
Size:
Universal
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-355-096 Universal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 17-355-096 Supplier National Safety Apparel™ Supplier No. S00CA0318
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High heat knit sleeve is inherently flame resistant and FR protection that will not wash or wear out.

  • Stretchable fabric for flexibility and easy range of motion
  • Thumbhole for added protection over the palm and back of hand
  • Knit fabric for comfortable fit
  • Lightweight and soft
  • Seamless design for no-rub comfort
  • Meets conductive heat level 2 at 285°F

Specifications

Product Type Welder Sleeve
Size Universal
Material OPF Blend
Color Black
Sterility Non-sterile
Style Standard Sleeve
Disposable Reusable
Closure Type Elastic Ends
For Use With (Application) Molten Metals, Welding
Certifications/Compliance ANSI/ISEA 105 Heat Level 2
Length (English) 18 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.