Learn More
Cytiva Carbon Capsule - 100g activated carbon (1/pkg)
Supplier:  Cytiva 12011
Encompass
Removes Odor, Colloids, and Organics from Liquids Using Activated Carbon-Finely divided carbon provides an active surface area of 70,000 m2-Ready to use and disposable This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3169