Learn More
Electron Microscopy Sciences Carbon Cohesive Sheet, Tech Grade Points
SDP
Catalog No. 5025627
Encompass_Preferred
These conductive cohesive sheets offer good adhesion to the specimen and yield good conductivity and background structures, even with particles as small as 15 microns. Produces minimum gas under vacuum conditions.
size: 5cm(W)x12cm(L)
Catalog No. 50-256-27 Supplier Electron Microscopy Sciences Supplier No. 7782201
These conductive cohesive sheets offer good adhesion to the specimen and yield good conductivity and background structures, even with particles as small as 15 microns. Produces minimum gas under vacuum conditions.
size: 5cm(W)x12cm(L)