Learn More
Ted Pella Inc Carbon Conductive Tape, 25mm W x 5m L
SDP
Catalog No. NC0041684
Encompass
Both sides of the 200µm thick conductive polycarbonate base have 30µm thick conductive adhesive. Total thickness is 260µm. Same material as the PELCO Image Tabs™. Protection for handling, storage and shipping is provided on one side by the 25µm thick, transparent liner and on the other side by the 40µm thick, white liner. The conductive adhesive is a carbon–filled acrylic adhesive, free of solvents. It can be removed from the specimen mount with ethyl acetate, ethanol, isopropanol or alcohols. Temperature maximum is 60°C (140°F). Small impurities of Cu, Si, Sb, S, Na, P and very small impurities of Fe and Mg can be found. Refrigeration will increase shelf life but a warm-up time of 1 hour is then required before use.
Catalog No. NC0041684 Supplier Ted Pella Inc Supplier No. 160848
Both sides of the 200µm thick conductive polycarbonate base have 30µm thick conductive adhesive. Total thickness is 260µm. Same material as the PELCO Image Tabs™. Protection for handling, storage and shipping is provided on one side by the 25µm thick, transparent liner and on the other side by the 40µm thick, white liner. The conductive adhesive is a carbon–filled acrylic adhesive, free of solvents. It can be removed from the specimen mount with ethyl acetate, ethanol, isopropanol or alcohols. Temperature maximum is 60°C (140°F). Small impurities of Cu, Si, Sb, S, Na, P and very small impurities of Fe and Mg can be found. Refrigeration will increase shelf life but a warm-up time of 1 hour is then required before use.