Learn More
Electron Microscopy Sciences Carbon Conductive Tape, Sheet Form
SDP
Catalog No. 50365852
Encompass_Preferred
EMS introduces this unique double sided carbon tape which is 260 microns in thickness. The base which is Polycarbonate and 200 microns thick has on both of its sides 30 microns of conductive glue giving it a total thickness of 260 microns. The tape has one transparent liner on one side and a white thicker one on the reverse which protects the tape in shipping. These are considered to be small impurities but they may be present
Width: 65 mm
Catalog No. 50-365-852 Supplier Electron Microscopy Sciences Supplier No. 7781965
EMS introduces this unique double sided carbon tape which is 260 microns in thickness. The base which is Polycarbonate and 200 microns thick has on both of its sides 30 microns of conductive glue giving it a total thickness of 260 microns. The tape has one transparent liner on one side and a white thicker one on the reverse which protects the tape in shipping. These are considered to be small impurities but they may be present
Width: 65 mm