Learn More
Ted Pella Inc ELCO Tabs™, Carbon Conductive Tabs, 25mm OD, 54/pk
SDP
Catalog No. NC9577655
Encompass
The simplicity of application and smooth, clean surfaces of PELCO Tabs™ Carbon Conductive Tabs are a significant improvement compared to many of the other common adhesives that have been used in SEM mounting. Both tab sides have a thick conductive adhesive (conductive inner tab is 35µm and the adhesive is 45µm on each side for a total thickness of 125µm [5 mils]) with a liner on both sides, a transparent liner and white liner respectively. They are ready for immediate use. The top protective liner does not have to be removed until the sample is ready to be mounted. This reduces possible contamination. Outgassing is negligible. The PELCO Tabs™ consist of a carbon fiber mat impregnated with a conductive adhesive. There is no internal carrier film. The conductive adhesive is a carbon filled acrylic, free of solvents. It can be removed from the specimen mount with ethyl acetate, ethanol, isopropanol, alcohols or our 3M™ Adhesive Remover 80924 which is solvent-free.
Catalog No. NC9577655 Supplier Ted Pella Inc Supplier No. 160842
The simplicity of application and smooth, clean surfaces of PELCO Tabs™ Carbon Conductive Tabs are a significant improvement compared to many of the other common adhesives that have been used in SEM mounting. Both tab sides have a thick conductive adhesive (conductive inner tab is 35µm and the adhesive is 45µm on each side for a total thickness of 125µm [5 mils]) with a liner on both sides, a transparent liner and white liner respectively. They are ready for immediate use. The top protective liner does not have to be removed until the sample is ready to be mounted. This reduces possible contamination. Outgassing is negligible. The PELCO Tabs™ consist of a carbon fiber mat impregnated with a conductive adhesive. There is no internal carrier film. The conductive adhesive is a carbon filled acrylic, free of solvents. It can be removed from the specimen mount with ethyl acetate, ethanol, isopropanol, alcohols or our 3M™ Adhesive Remover 80924 which is solvent-free.