Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cambridge Isotope Laboratories Carbon dioxide (18O2 95%) 1 L

Catalog No. 5000166507
Encompass

Carbon dioxide (18O2 95%) 1 L

Catalog No. 50-001-66507 Supplier Cambridge Isotope Laboratories Supplier No. OLM1861LB
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Carbon dioxide (18O2 95%) 1 L

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.