Learn More
Crystal Chem Inc Carbon Dioxide (CO2) Assay Kit, 96-tests
SDP
Catalog No. 501947667
Encompass_Preferred
The Carbon Dioxide (CO2) Assay Kit is used to quantify levels of carbon dioxide in serum and plasma using only a 3 µL sample volume. Highlights: Almost no interference from triglycerides, ascorbate, bilirubin and hemolysis. Provides exceptional performance at a reasonable price. Results in as little as five minutes. Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7667 Supplier Crystal Chem Inc Supplier No. 80139
The Carbon Dioxide (CO2) Assay Kit is used to quantify levels of carbon dioxide in serum and plasma using only a 3 µL sample volume. Highlights: Almost no interference from triglycerides, ascorbate, bilirubin and hemolysis. Provides exceptional performance at a reasonable price. Results in as little as five minutes. Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.