Learn More
VAISALA INC CO2 Probe
Catalog No. NC1653955
Encompass_Preferred
The GMP251 is a all-in-one probe, which makes it easy and fast to install. The probe is resistant to dust and most chemicals, such as, H2O2 and alcohol-based cleaning agents. The probe's surface is smooth, which makes it easy to clean. With the wide operating temperature range, the GMP251 is suitable for a large number of applications. The probe provides accurate and stable measurements also in varying conditions.
Catalog No. NC1653955 Supplier VAISALA INC Supplier No. GMP251
The GMP251 is a all-in-one probe, which makes it easy and fast to install. The probe is resistant to dust and most chemicals, such as, H2O2 and alcohol-based cleaning agents. The probe's surface is smooth, which makes it easy to clean. With the wide operating temperature range, the GMP251 is suitable for a large number of applications. The probe provides accurate and stable measurements also in varying conditions.