Learn More
MilliporeSigma™ Supelco™ Carbon Dioxide Trap
Catalog No. 111022612 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
100 cc, Fitting: 1/4 in.
100 cc, Fitting: 1/8 in.
250 cc, Fitting: 1/8 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
11-102-2612 100 cc, Fitting: 1/8 in.
11-102-2614 250 cc, Fitting: 1/8 in.
11-102-2613 100 cc, Fitting: 1/4 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-102-2612 Supplier MilliporeSigma™ Supelco™ Supplier No. 503185
Add to Cart
Add to Cart
As carbon dioxide is absorbed, the greenish-brown material turns white due to of the formation of sodium carbonate. The trap body is constructed of borosilicate glass, and the fittings are nickel-plated with sintered stainless steel frits.
  • Sodium hydroxide nonfibrous silicate material is universally accepted for its high absorptive capacity and indicating properties for carbon dioxide
  • Typically, this material will absorb 20-30% of its weight in carbon dioxide before replacement of the saturated material is required
  • A moisture trap should be installed downstream from this unit to absorb the moisture that is produced
  • Exercise extreme caution when refilling the trap due to the caustic nature of the absorbent