Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbon disulfide, 99.9%, for HPLC

Catalog No. AC445660025
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC445660025 2.5 L Glass Bottle
AC445661000 100 mL Glass Bottle
AC445660010 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC445660025 Supplier Thermo Scientific Chemicals Supplier No. 445660025
  • $453.50
    $385.65 / Each of 1

    Save $67.85

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 75-15- | CS2 | 76.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-15-
Molecular Formula CS2
Molecular Weight (g/mol) 76.13

Specifications

Melting Point -111.0°C
Density 1.2620g/mL
Quantity 2.5 L
Flash Point −30°C
Packaging Glass Bottle
Refractive Index 1.627
Assay Percent Range 99.85% min. (GC)
Specific Gravity 1.262
Linear Formula CS2
Viscosity 0.363 mPa.s (20°C)
Merck Index 151811
Boiling Point 46.0°C
Solubility Information Solubility in water: 2.9g/L (20°C). Other solubilities: miscible with anhydrous methanol,ethanol,ether,,benzene,chloroform,carbon tetrachloride and oils
Formula Weight 76.13
Percent Purity 99.9%
Grade HPLC
Chemical Name or Material Carbon disulfide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Causes damage to organs through prolonged or repeated exposure.
Suspected of damaging fertility.
Suspected of damaging the unborn child.
Highly fla
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
EINECSNumber 200-843-6

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.