Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbon disulfide, ≥99.9%, Reagent Grade, Solstice

Catalog No. 60046858 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-858 100 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-046-858 Supplier Solstice Supplier No. 424641100ML

missing translation for 'validMainframeMsg'

Reagent Grade, purified by redistillation,≥99.9%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-15-0
Molecular Formula CS2
Molecular Weight (g/mol) 76.131
MDL Number MFCD00011321
InChI Key QGJOPFRUJISHPQ-UHFFFAOYSA-N
Synonym carbon disulfide, carbon disulphide, carbon bisulfide, dithiocarbonic anhydride, carbon bisulphide, schwefelkohlenstoff, weeviltox, wegla dwusiarczek, solfuro di carbonio, carbondisulphide
PubChem CID 6348
ChEBI CHEBI:23012
IUPAC Name methanedithione
SMILES C(=S)=S

Specifications

Density 1.266g/mL at 25°C
Quantity 100 mL
Explosion Limit 0.5
Flash Point −30°C (−22°F)
Ignition Point 212°F
Packaging Glass Bottle
UN Number UN1131
Beilstein 1098293
Formula Weight 76.14g/mol
Percent Purity ≥99.9%
Grade ReagentPlus
Chemical Name or Material Carbon disulfide, Purified by redistillation
Show More Show Less
Hazard Statement P201-P210-P261-P280-P304 + P340 + P312-P308 + P313
DOTInformation class 3 - PG 1 - Carbon disulfide
RTECSNumber 200-843-6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.