Learn More
Sigma Aldrich Fine Chemicals Biosciences Carbon disulfide ACS reage1L
Catalog No. 502899196
Encompass
Carbon disulfide is a highly volatile flammable liquid with low ignition temperature. It can be synthesized by reacting hydrocarbon gas with sulfur. It is an important raw material for preparing viscose rayon and cellophane film.
Catalog No. 50-289-9196 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 1801731L
Carbon disulfide is a highly volatile flammable liquid with low ignition temperature. It can be synthesized by reacting hydrocarbon gas with sulfur. It is an important raw material for preparing viscose rayon and cellophane film.