Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbon Disulfide, ACS Reagent, ≥99.9%, Solstice

Catalog No. 60045026 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-026 1 L Glass Bottle
60-045-027 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-045-026 Supplier Solstice Supplier No. 1801731L
Add to Cart
Add to Cart

ACS reagent,≥99.9%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-15-0
Molecular Formula CS2
Molecular Weight (g/mol) 76.131
MDL Number MFCD00011321
InChI Key QGJOPFRUJISHPQ-UHFFFAOYSA-N
Synonym carbon disulfide, carbon disulphide, carbon bisulfide, dithiocarbonic anhydride, carbon bisulphide, schwefelkohlenstoff, weeviltox, wegla dwusiarczek, solfuro di carbonio, carbondisulphide
PubChem CID 6348
ChEBI CHEBI:23012
IUPAC Name methanedithione
SMILES C(=S)=S

Specifications

Melting Point -76°C
Density 1.266g/mL at 25°C
Quantity 1 L
Explosion Limit 0.5
Flash Point −30°C (−22°F)
Ignition Point 212°F
Packaging Glass Bottle
Physical Form Liquid
Refractive Index n20/D 1.627(lit.)
UN Number UN1131
Beilstein 1098293
Boiling Point 46°C
Formula Weight 76.14g/mol
Percent Purity ≥99.9%
Grade ACS
Chemical Name or Material Carbon Disulfide
Show More Show Less
Hazard Statement P210-P260-P280-P305 + P351 + P338-P370 + P378-P403 + P235
Precautionary Statement H225-H315-H319-H332-H361fd-H372
DOTInformation Class 3 - PG 1 - Carbon disulfide
EINECSNumber 200-843-6
RTECSNumber FF6650000
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.