Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbon disulfide, CHROMASOLV™, for HPLC, ≥99.9%, Solstice

Catalog No. 6000396 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
1 L
4 x 2 L
6 x 1 L
Packaging:
Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-003-96 6 x 1 L Glass Bottle
60-003-94 1 L Glass Bottle
60-003-93 100 mL Glass Bottle
60-003-95 4 x 2 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-003-96 Supplier Solstice Supplier No. 2706606X1L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CHROMASOLV™, for HPLC,≥99.9%

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-15-0
Molecular Formula CS2
Molecular Weight (g/mol) 76.131
MDL Number MFCD00011321
InChI Key QGJOPFRUJISHPQ-UHFFFAOYSA-N
Synonym carbon disulfide, carbon disulphide, carbon bisulfide, dithiocarbonic anhydride, carbon bisulphide, schwefelkohlenstoff, weeviltox, wegla dwusiarczek, solfuro di carbonio, carbondisulphide
PubChem CID 6348
ChEBI CHEBI:23012
IUPAC Name methanedithione
SMILES C(=S)=S

Specifications

Melting Point -112°C to -111°C (literature)
Absorbance max. 0.50 at 388 nm, max. 1.0 at 386 nm, max. 0.01 at 750 nm, max. 0.01 500 nm, max. 0.05 at 410 nm, max. 0.10 at 403 nm, max. 0.25 at 394 nm
CAS Max % 100.0000%
Density 1.266 g/mL at 25 °C(lit.)
Quantity 6 x 1 L
Flash Point −30°C
Packaging Glass Bottle
Linear Formula CS2
UN Number UN1131
Beilstein 1098293
Boiling Point 46°C
Formula Weight 76.14
Percent Purity >99.9%
Grade HPLC
Chemical Name or Material Carbon disulfide, CHROMASOLV™, for HPLC, ≥99.9%
Show More Show Less
ShelfLife 1260 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: I; Proper Shipping Name: Carbon disulfide
EINECSNumber 200-843-6
Recommended Storage Room Temp
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.