Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbon Disulfide, Solstice

Catalog No. 60048399 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-048-399 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-048-399 Supplier Solstice Supplier No. 316271L
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

for IR spectroscopy, Puriss. p.a., ACS reagent, Reag. Ph. Eur.,≥99.9% (GC)

  • Solstice has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-15-0
Molecular Formula CS2
Molecular Weight (g/mol) 76.131
MDL Number MFCD00011321
InChI Key QGJOPFRUJISHPQ-UHFFFAOYSA-N
Synonym carbon disulfide, carbon disulphide, carbon bisulfide, dithiocarbonic anhydride, carbon bisulphide, schwefelkohlenstoff, weeviltox, wegla dwusiarczek, solfuro di carbonio, carbondisulphide
PubChem CID 6348
ChEBI CHEBI:23012
IUPAC Name methanedithione
SMILES C(=S)=S

Specifications

Density 1.266g/mL at 25°C
Quantity 1 L
Explosion Limit 0.5
Flash Point −30°C (−22°F)
Ignition Point 212°F
UN Number 1131
Beilstein 1098293
Formula Weight 76.14g/mol
Percent Purity ≥99.9% (GC)
Grade puriss. p.a.
Chemical Name or Material Carbon Disulfide, Reag. Ph. Eur.
Show More Show Less
Hazard Category Danger
Hazard Statement P201-P210-P261-P280-P304 + P340 + P312-P308 + P313
Precautionary Statement H225-H315-H319-H332-H361fd-H372
EINECSNumber 200-843-6
RTECSNumber FF6650000
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.