Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbon Disulfide, ULTRA RESI-ANALYZED™, J.T. Baker™

Catalog No. 02002801 Shop All Avantor Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Clear PVC-coated Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-002-801 500 mL Clear PVC-coated Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-002-801 Supplier Avantor Supplier No. E35001
 Restricted Item
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

High purity solvents and salts developed for organic residue extraction/concentration procedures

    For use in Gas Chromatography (GC), Suitable for NIOSH Analytical Methods 1003, 1500, & 1501 for Hydrocarbon Analysis and Spectroscopy; For Laboratory, Research, or Manufacturing Use

Chemical Identifiers

CAS 75-15-0
Molecular Formula CS2
Molecular Weight (g/mol) 76.131
InChI Key QGJOPFRUJISHPQ-UHFFFAOYSA-N
Synonym carbon disulfide, carbon disulphide, carbon bisulfide, dithiocarbonic anhydride, carbon bisulphide, schwefelkohlenstoff, weeviltox, wegla dwusiarczek, solfuro di carbonio, carbondisulphide
PubChem CID 6348
ChEBI CHEBI:23012
IUPAC Name methanedithione
SMILES C(=S)=S

Specifications

Quantity 500 mL
Flash Point −30.00°C
Packaging Clear PVC-coated Glass Bottle
Specific Gravity 1L = 1.26kg
Formula Weight 76.13
Grade ULTRA RESI-ANALYZED
Chemical Name or Material Carbon Disulfide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.