Learn More
Sigma Aldrich Fine Chemicals Biosciences Carbon dsulfde ACS reage500ML
Catalog No. 502899291
Encompass
Carbon disulfide is a highly volatile flammable liquid with low ignition temperature. It can be synthesized by reacting hydrocarbon gas with sulfur. It is an important raw material for preparing viscose rayon and cellophane film.
Catalog No. 50-289-9291 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 180173500ML
Carbon disulfide is a highly volatile flammable liquid with low ignition temperature. It can be synthesized by reacting hydrocarbon gas with sulfur. It is an important raw material for preparing viscose rayon and cellophane film.