Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbon felt, 1.27cm (0.5 in.) thick, 95%, Thermo Scientific™

Catalog No. AA43026RR
Change view
Click to view available options
Quantity:
10 x 10 cm
30 x 30 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43026RR 10 x 10 cm
AA43026RF 30 x 30 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA43026RR Supplier Thermo Scientific Chemicals Supplier No. 43026RR

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00133992

Specifications

Quantity 10 x 10 cm
Percent Purity 95%
Chemical Name or Material Carbon felt, 1.27cm (0.5 in.) thick
EINECSNumber 231-955-3
TSCA U
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.