Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbon felt, 3.18mm (0.125in) thick, 99.0%, Thermo Scientific Chemicals

Catalog No. AA43199RR
Change view
Click to view available options
Quantity:
10 x 10 cm
25 x 25 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43199RR 10 x 10 cm
AA43199RY 25 x 25 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA43199RR Supplier Thermo Scientific Chemicals Supplier No. 043199RR
Add to Cart

Typical thermal conductivity of 1.75 BTU in/hr/ft²/°F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00133992

Specifications

Assay Percent Range 99.0%
Quantity 10 x 10 cm
Chemical Name or Material Carbon felt, Processed at 1400°C
EINECSNumber 231-153-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.