Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbon felt, 6.35mm (0.25in) thick, 99.0%, Thermo Scientific Chemicals

Catalog No. AA43200RF
Change view
Click to view available options
Quantity:
10 x 10 cm
30 x 30 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43200RF 30 x 30 cm
AA43200RR 10 x 10 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA43200RF Supplier Thermo Scientific Chemicals Supplier No. 043200RF
Only null left
Add to Cart

Typical thermal conductivity of 1.75 BTU in/hr/ft²/°F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1333-86-4
Molecular Formula C
Molecular Weight (g/mol) 12.01
MDL Number MFCD00133992
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N

Specifications

Density 2.26g/cm3
Assay Percent Range 99.0%
Quantity 30 x 30 cm
Chemical Name or Material Carbon
EINECSNumber 231-955-3
TSCA U
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.