Learn More
Electron Microscopy Sciences EMS Carbon Fiber Cord Standard Purity, Fine Strands 100 MT
SDP
Catalog No. 5024880
Encompass
Sometimes referred to as 'braid', carbon fiber consists of eight separate strands of individual carbon fiber braided together. The fiber is manufactured so that, when cut, the ends resist unravelling.
Catalog No. 50-248-80 Supplier Electron Microscopy Sciences Supplier No. 91046
Sometimes referred to as 'braid', carbon fiber consists of eight separate strands of individual carbon fiber braided together. The fiber is manufactured so that, when cut, the ends resist unravelling.