Learn More
Electron Microscopy Sciences EMS Carbon Fiber Cord Standard Grade 10 MT
SDP
Catalog No. 501900136
Encompass_Preferred
Sometimes referred to as 'braid', carbon fiber consists of eight separate strands of individual carbon fiber braided together. The fiber is manufactured so that, when cut, the ends resist unravelling.
Catalog No. 50-190-0136 Supplier Electron Microscopy Sciences Supplier No. 9104610
Sometimes referred to as 'braid', carbon fiber consists of eight separate strands of individual carbon fiber braided together. The fiber is manufactured so that, when cut, the ends resist unravelling.