Learn More
Electron Microscopy Sciences EMS Carbon Fiber Cord Standard Grade 1 MT
SDP
Catalog No. 501900135
Encompass_Preferred
Sometimes referred to as 'braid', carbon fiber consists of eight separate strands of individual carbon fiber braided together. The fiber is manufactured so that, when cut, the ends resist unravelling.
Catalog No. 50-190-0135 Supplier Electron Microscopy Sciences Supplier No. 910461
Sometimes referred to as 'braid', carbon fiber consists of eight separate strands of individual carbon fiber braided together. The fiber is manufactured so that, when cut, the ends resist unravelling.