Learn More
World Precision Instrument Carbon Fiber Electrode, Dia 10 um,Length 250 um
SDP
Catalog No. 502521202
Encompass
Electrochemical detection of oxidizable compounds
Carbon fiber microelectrodes have been used in both the detection of oxidizable compounds (Gonon, et al., 1978; Cahill and Wightman, 1995) and extracellular single-unit recording (Armstrong-James and Millar, 1979). WPI's ultra-sensitive and low-noise carbon fiber (CF) electrodes can be applied, with our MicroC Potentiostat or similar instruments, in the electrochemical detection of catecholamines (epinephrine, norepinephrine and dopamine), indolamines (serotonin, 5-HT or melatonin), Fe (II) and other oxidizable compounds.
Catalog No. 50-252-1202 Supplier World Precision Instrument Supplier No. CFN10250
Electrochemical detection of oxidizable compounds
Carbon fiber microelectrodes have been used in both the detection of oxidizable compounds (Gonon, et al., 1978; Cahill and Wightman, 1995) and extracellular single-unit recording (Armstrong-James and Millar, 1979). WPI's ultra-sensitive and low-noise carbon fiber (CF) electrodes can be applied, with our MicroC Potentiostat or similar instruments, in the electrochemical detection of catecholamines (epinephrine, norepinephrine and dopamine), indolamines (serotonin, 5-HT or melatonin), Fe (II) and other oxidizable compounds.