Learn More
World Precision Instrument Carbon Fiber Electrode, Dia 30 um, L 100 um
SDP
Catalog No. 502521201
Encompass
Electrochemical detection of oxidizable compounds
Carbon fiber microelectrodes have been used in both the detection of oxidizable compounds (Gonon, et al., 1978; Cahill and Wightman, 1995) and extracellular single-unit recording (Armstrong-James and Millar, 1979). WPI™s ultra-sensitive and low-noise carbon fiber (CF) electrodes can be applied, with our Micro-C Potentiostat or similar instruments, in the electrochemical detection of catecholamines (epinephrine, norepinephrine and dopamine), indolamines (serotonin, 5-HT or melatonin), Fe (II) and other oxidizable compounds.
Catalog No. 50-252-1201 Supplier World Precision Instrument Supplier No. CFN30100
Electrochemical detection of oxidizable compounds
Carbon fiber microelectrodes have been used in both the detection of oxidizable compounds (Gonon, et al., 1978; Cahill and Wightman, 1995) and extracellular single-unit recording (Armstrong-James and Millar, 1979). WPI™s ultra-sensitive and low-noise carbon fiber (CF) electrodes can be applied, with our Micro-C Potentiostat or similar instruments, in the electrochemical detection of catecholamines (epinephrine, norepinephrine and dopamine), indolamines (serotonin, 5-HT or melatonin), Fe (II) and other oxidizable compounds.