Learn More
World Precision Instrument Carbon Fiber Electrode, Length 100 um
SDP
Catalog No. 502538903
Encompass
Electrochemical detection of oxidizable compounds
Carbon fiber microelectrodes have been used in both the detection of oxidizable compounds (Gonon, et al., 1978; Cahill and Wightman, 1995) and extracellular single-unit recording (Armstrong-James and Millar, 1979). WPI™s ultra-sensitive and low-noise carbon fiber (CF) electrodes can be applied, with our Micro-C Potentiostat or similar instruments, in the electrochemical detection of catecholamines (epinephrine, norepinephrine and dopamine), indolamines (serotonin, 5-HT or melatonin), Fe (II) and other oxidizable compounds.
Catalog No. 50-253-8903 Supplier World Precision Instrument Supplier No. CF30100
Electrochemical detection of oxidizable compounds
Carbon fiber microelectrodes have been used in both the detection of oxidizable compounds (Gonon, et al., 1978; Cahill and Wightman, 1995) and extracellular single-unit recording (Armstrong-James and Millar, 1979). WPI™s ultra-sensitive and low-noise carbon fiber (CF) electrodes can be applied, with our Micro-C Potentiostat or similar instruments, in the electrochemical detection of catecholamines (epinephrine, norepinephrine and dopamine), indolamines (serotonin, 5-HT or melatonin), Fe (II) and other oxidizable compounds.