Learn More
Elemental Scientific Inc CARBON FIBER ESI SC-DX PROBE
SDP
Catalog No. NC2256668
Encompass_Preferred
CARBON FIBER ESI SC-DX PROBE (ORANGE MARKER) FOR ST NEBULIZERS - 150CM PAILLARY LENGTH. 0.50MM ID
This item is not returnable. View return policy
Catalog No. NC2256668 Supplier Elemental Scientific Inc Supplier No. ES50373505150
CARBON FIBER ESI SC-DX PROBE (ORANGE MARKER) FOR ST NEBULIZERS - 150CM PAILLARY LENGTH. 0.50MM ID