Learn More
Mopec Carbon Fiber Latent Print Brush
SDP
Catalog No. 22446334 Shop All Mopec Products
Change view
Click to view available options
For Use With (Application):
Latent Fingerprint Development
Catalog No. For Use With (Application)
22-446-334 Latent Fingerprint Development
1 options
Catalog No. 22-446-334 Supplier Mopec Supplier No. FS347
Add to Cart
Add to Cart
Fine carbon fiber bristles support development of latent fingerprints while helping minimize damage to evidence. A sturdy plastic handle provides controlled use, and an anti-static storage tube protects the bristles between applications for organized handling and storage.
Construction
  • Bristles made from carbon fibers
  • Anti-static tube
  • Sturdy plastic handle
  • Low friction quality
  • Corrosion and moisture-resistant
Dimensions
  • Handle: 4.13 in. L
  • Carbon filament: 2.16 in. L
Materials
  • Carbon fiber
  • Plastic
Specifications
Product Type Latent Print Brush
For Use With (Application) Latent Fingerprint Development