Learn More
Perkin Elmer US LLC Carbon Fiber Sample Probe, Blue Band, 0.5 mm I.D.
Catalog No. 502105275
Encompass_Preferred
The carbon fiber reinforced probes are designed to be mounted to a Z-drive with two probe clamps andwithout a guide plate. The carbon fiber probes can also be used with the prior version Z-drives, but theguide plate must be used. All wetted surfaces (liquid contact) of both the carbon fiber and Ultem probes are inert and identical(PTFE). Color bands indicate inside diameters and are the same for both probe types.
Catalog No. 50-210-5275 Supplier Perkin Elmer US LLC Supplier No. N0777547
The carbon fiber reinforced probes are designed to be mounted to a Z-drive with two probe clamps andwithout a guide plate. The carbon fiber probes can also be used with the prior version Z-drives, but theguide plate must be used. All wetted surfaces (liquid contact) of both the carbon fiber and Ultem probes are inert and identical(PTFE). Color bands indicate inside diameters and are the same for both probe types.