Learn More
Electron Microscopy Sciences CARBON FIBRE fine strands (1m)
SDP
Catalog No. 5024879
Encompass
Sometimes referred to as 'braid', carbon fibre consists of eight separate strands of individual carbon fibre braided together. The fibre is manufactured so that, when cut, the ends resist unravelling. The fibre density is about 1.6g/m. Carbon Fibre fine strands are convenient to use and result in thin to medium coating using a single strand or several lengths.
Catalog No. 50-248-79 Supplier Electron Microscopy Sciences Supplier No. 91045
Sometimes referred to as 'braid', carbon fibre consists of eight separate strands of individual carbon fibre braided together. The fibre is manufactured so that, when cut, the ends resist unravelling. The fibre density is about 1.6g/m. Carbon Fibre fine strands are convenient to use and result in thin to medium coating using a single strand or several lengths.