Learn More
Electron Microscopy Sciences EMS Style 2A Carbon Fiber Replacement Tips, OAL
SDP
Catalog No. 501930869
Encompass_Preferred
Durable self-aligning replacement carbon fiber tips and screws for replaceable tip tweezers Style 2A. Flat round tips.
Material: Carbon Fiber
Catalog No. 50-193-0869 Supplier Electron Microscopy Sciences Supplier No. 7832557
Durable self-aligning replacement carbon fiber tips and screws for replaceable tip tweezers Style 2A. Flat round tips.
Material: Carbon Fiber