Learn More
Electron Microscopy Sciences EMS Style 7 Carbon Fiber Replacement Tips, OAL
SDP
Catalog No. 501930878
Encompass_Preferred
Durable self-aligning replacement carbon fiber tips and screws for replaceable tip tweezers Style 7. Very fine, curved tips.
Material: Carbon Fiber
Catalog No. 50-193-0878 Supplier Electron Microscopy Sciences Supplier No. 7832566
Durable self-aligning replacement carbon fiber tips and screws for replaceable tip tweezers Style 7. Very fine, curved tips.
Material: Carbon Fiber