Learn More
Perkin Elmer US LLC Carbon Fiber Support Probe, 0.25 mm I.D.
Catalog No. 502112218
Encompass_Preferred
This Carbon fiber support SC-autosampler probe for ST nebulizers, 0.25 mm I.D. 100 µL/min uptake rate, 150 cm capillary.
Catalog No. 50-211-2218 Supplier Perkin Elmer US LLC Supplier No. N0777826
This Carbon fiber support SC-autosampler probe for ST nebulizers, 0.25 mm I.D. 100 µL/min uptake rate, 150 cm capillary.