Learn More
Valutek™ Carbon Fiber Ultrasonic Seal Standard Weight Polyester Wiper
Catalog No. 191502147
Change view
Click to view available options
Dimensions (L x W):
9 x 9 in.
Catalog No. Dimensions (L x W)
19-150-2147 9 x 9 in.
1 options
This item is not returnable. View return policy
Catalog No. 19-150-2147 Supplier Valutek™ Supplier No. VTPNWESD99
Add to Cart
Add to Cart
This item is not returnable. View return policy
Constructed from 100% continuous-filament polyester in a double-knit, no-run, interlock pattern with a conductive fiber designed to dissipate electrical charge. This substrate exhibits moderate absorbency and low levels of particulate and extractable counts.
  • 100% continuous filament, double-knit polyester fiber
  • Cold knife cut edges
  • Chemically compatible with IPA and other common solvents
  • Excellent absorbency and abrasion resistance
  • Low levels of particulate and extractable counts
  • Low in fibers generation
  • Laundered and packaged in a controlled environment, all Valutek™ wipers are tested and manufactured in ISO compliant facilities under Valutek™ inspection and strict process control to ensure Valutek™ quality standard and product specifications
  • This product is recommended for use in a cleanroom Class 100-1,000 (ISO 5-6) critical environment
  • It is commonly used in a wide variety of applications, such as optics and LCD screen cleaning, electronic manufacturing, optical cleaning and pharmaceutical manufacturing
Specifications
Material Polyester
Dimensions (L x W) 9 x 9 in.
Sterility Non-sterile