Learn More
Electron Microscopy Sciences CARBON FIBRE fine strands (100m)
SDP
Catalog No. 501901841
Encompass_Preferred
Sometimes referred to as 'braid', carbon fibre consists of eight separate strands of individual carbon fibre braided together. The fibre is manufactured so that, when cut, the ends resist unravelling. The fibre density is about 1.6g/m. Carbon Fibre fine strands are convenient to use and result in thin to medium coating using a single strand or several lengths.
Catalog No. 50-190-1841 Supplier Electron Microscopy Sciences Supplier No. 91045100
Sometimes referred to as 'braid', carbon fibre consists of eight separate strands of individual carbon fibre braided together. The fibre is manufactured so that, when cut, the ends resist unravelling. The fibre density is about 1.6g/m. Carbon Fibre fine strands are convenient to use and result in thin to medium coating using a single strand or several lengths.