Learn More
Ted Pella Inc Carbon Film only on 200 mesh, Copper, 50/pk
SDP
Catalog No. NC0452762
Encompass
Carbon films with a thickness of 15-25nm with no Formvar used during manufacturing. Completely free of Formvar. Carbon films are thin and highly transparent to electrons. They exhibit very fine grain and minimal interference with specimen structure. Films are ready to use upon receipt. The pure carbon films tend to be more delicate than support films with a Formvar backing and require more delicate handling during specimen preparation.
Catalog No. NC0452762 Supplier Ted Pella Inc Supplier No. 01840F
Carbon films with a thickness of 15-25nm with no Formvar used during manufacturing. Completely free of Formvar. Carbon films are thin and highly transparent to electrons. They exhibit very fine grain and minimal interference with specimen structure. Films are ready to use upon receipt. The pure carbon films tend to be more delicate than support films with a Formvar backing and require more delicate handling during specimen preparation.