Learn More
Electron Microscopy Sciences Carbon Support Film 5-6 nm thick on Square 400 mesh Copper Grid
SDP
Catalog No. 5024891
Encompass_Preferred
A thin film of pure carbon. All of our coated grids are optically checked followed by batch testing in the EM. Packed in grid storage box. NOTE: All of our film is laid on the shiny side of the grid. Standard thickness is 5-6 nm.
Grid: Copper
Catalog No. 50-248-91 Supplier Electron Microscopy Sciences Supplier No. CF400CU50
A thin film of pure carbon. All of our coated grids are optically checked followed by batch testing in the EM. Packed in grid storage box. NOTE: All of our film is laid on the shiny side of the grid. Standard thickness is 5-6 nm.
Grid: Copper