Learn More
Strem, An Ascensus Company CAS# 308068-56-6. 1g. Carbon nanotubes, multi-walled (diameter = ~140nm, length = ~7 microns) (>90% nanotubes). MFCD00133992
SDP
Catalog No. 5090108799
Encompass
CAS# 308068-56-6. 1g. Carbon nanotubes, multi-walled (diameter = ~140nm, length = ~7 microns) (>90% nanotubes). MFCD00133992. Molecular Weight: . Molecular Formula: . Color/form: black pwdr. Strem# 06-0470. http://www.strem.com/catalog/v/06-0470/
Catalog No. 50-901-08799 Supplier Strem, An Ascensus Company Supplier No. 0604701G
CAS# 308068-56-6. 1g. Carbon nanotubes, multi-walled (diameter = ~140nm, length = ~7 microns) (>90% nanotubes). MFCD00133992. Molecular Weight: . Molecular Formula: . Color/form: black pwdr. Strem# 06-0470. http://www.strem.com/catalog/v/06-0470/