Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Carbon nanotube, few-walled

Catalog No. 900788250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-078-8250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-078-8250MG Supplier Sigma Aldrich Supplier No. 900788250MG

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Signis(R) FW100 is engineered to meet the growing demand for a material with the high conductivity, large surface area, and to contain mostly double and triple walled carbon nanotubes. Applications for few-walled carbon nanotubes include advanced battery electrode additives, super/ultracapacitor materials, and more.Battery materials.Supplementation of lithium sulphur battery cathode materials.Signis is a registered trademark of Chasm Advanced Materials

Specifications

CAS 308068-56-6
Quantity 250 mg
Synonym FWCNT; FWNT; Few wall carbon nanotubes; Few-walled carbon nanotubes; Signis™ FW100
Percent Purity ≥94% (carbon as CNT by TGA)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.