Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Carbon nanotube, single-walled, carboxylic acid functionalized

Catalog No. 652490250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
65-249-0250MG 250 mg
65-249-01G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 65-249-0250MG Supplier Sigma Aldrich Supplier No. 652490250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Single walled nanotubes were prepared by the electric arc discharge method. The SWNTs were functionalized by purifying them in nitric acid. The carboxylic acid (-COOH) functionality along the length of the nanotube can be derivatized with a variety of functional groups. The average diameter of the nanotubes is 1.4nm +/- 0.1nm. Transport properties of the SWNTs in soil was studied.Carboxylated SWNTs were used to modify an indium tin oxide (ITO) electrode towards the fabrication of a novel electrochemical immunosensor. Carboxylated SWNTs was further multifunctionalized and were used in studying the influence of attached chemical group on the sorption of pyrene conjugates of oligonucleotides. An electrochemical DNA biosensor was reportedly designed using functionalized SWNTs. It was used in the preparation of the chlorocarbonyl-functionalized SWNT. It was used to fabricate SWNTs/(Pb, Zn)-phosphate glass composite structure for the dual use as light sensor and photocurrent converter. Interaction of carboxylated SWNT with Caco-2 was investigated in detail.

Specifications

Quantity 250 mg
Synonym SWNT, carboxylic acid functionalized
Percent Purity >90%carbon basis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.