Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Carbon nanotube, single-walled, conductive aqueous ink, SWCNT 0.2 mg/mL

Catalog No. 791490100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
79-149-0100ML 100 mL
79-149-025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 79-149-0100ML Supplier Sigma Aldrich Supplier No. 791490100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

This conductive ink is aqueous based and contains highly conductive single wall carbon nanotubes and is formulated for spray coating.This ink is primarily intended for making highly transparent conductive films on a variety of substrates.Product of Chasm Advanced Materials, Inc.Signis is a registered trademark of Chasm Advanced Materials, Inc.CoMoCAT is a trademark of Chasm Advanced Materials, Inc.CHASM is a trademark of Chasm Advanced Materials, Inc.

Specifications

Quantity 100 mL
Synonym CG300-Aqueous Ink; SWCNT Ink; SWNT Ink; SWeNT AC100; Single-Walled Carbon Nanotube Ink
Density 1 g/cm3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.