Learn More
Sigma Aldrich Carbon nanotube, single-walled, solvent-based conductive ink, SWCNT
Catalog No. 79246225ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mL
Catalog No. Quantity
79-246-225ML 25 mL
1 options
Catalog No. 79-246-225ML Supplier Sigma Aldrich Supplier No. 79246225ML
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Single walled carbon nanotubes offer an alternative to metallic conductive inks. The nanotubes exhibit high conductivity, oxidation resistance. Carbon nanomaterial based conductive inks are used in printed and flexible electronics, thin film transistors, electrochemical sensors, supercapacitors and photovoltaics.This solvent based conductive ink is primarily formulated for Screen Printing Transparent ConductorsProduct of Chasm Advanced Materials, Inc.Signis is a registered trademark of Chasm Advanced Materials, Inc.CoMoCAT is a trademark of Chasm Advanced Materials, Inc.CHASM is a trademark of Chasm Advanced Materials, Inc.
Specifications
Quantity 25 mL
Synonym SWCNT Ink; SWNT Ink; SWeNT V2V ink; SWeNT VC101 ink; Single-Walled Carbon Nanotube Ink; conductive ink