Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Carbon nanotube, single-walled, polyaminobenzene sulfonic acid functionalized

Catalog No. 502907134
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-290-7134 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-290-7134 Supplier Sigma Aldrich Supplier No. 639230100MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Average diameter of individual SWCNT = 1.4nm +/- 0.1nmMethod for dispersing in water: In solid form SWNTs are highly entangled and associated in macroscopic particles. It requires significant effort to break such an association and bring the material in solution. Sonicate 50 mg of material in 2 ml water for 30 min. Add 8 ml of water and sonicate for additional 90 min. Typically, after this procedure all material is completely dispersed according to visual observation. For quantitative measurement of the solubility, allow the dispersion to stand overnight at room temperature. Because of the high optical density of concentrated dispersion, dilute an aliquot from the upper layer of the studied dispersion by a factor of 100. The concentration of the diluted solution is estimated from the recorded NIR spectra using a plot of the absorbance at 9750 cm-1 compared to the concentration of a standard solution (see reference).
TRUSTED_SUSTAINABILITY

Specifications

MDL Number MFCD05865412
Quantity 100 mg
Synonym Carbon nanotube, single-walled, PABS functionalized; SWNT, PABS functionalized
Molecular Weight (g/mol) 400-600
Percent Purity 75-85%carbon basis
Biological Activity Respiratory system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.