Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Carbon Paper Disc
GREENER_CHOICESCI_ED

Catalog No. S139427 Shop All Supertek Scientific Products
Change view
Click to view available options
No. per Pack:
100/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Pack
S139427 100/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S139427 Supplier Supertek™ Supplier No. PH10552
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

43 mm carbon paper disc.

  • Carbon Paper Disc
  • Non-sterile
  • 43 mm Diameter

Specifications

Autoclavable Not Autoclavable
Disposable Single-use
Filter Type Carbon Paper
Format Disc
Material Carbon Paper
Material (Membrane) Paper
Pore Size None
Sterility Non-sterile
No. per Pack 100/Cs.
Type Carbon Paper Disc
For Use With (Equipment) Acceleration Timer
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.